5-Fluoro-2-methylphenylboronic acid 99%

Warning

Synonyms: 5-Fluoro-2-methylbenzeneboronic acid2-Methyl-5-fluorophenylboronic acid

L19819-03 L19819-06 L19819-06 L19819-03
CAAAAL19819-06EA 292.98 CAD
CAAAAL19819-06 AAAL19819-06 CAAAAL19819-03 AAAL19819-03
5-Fluoro-2-methylphenylboronic acid 99%
5-Fluoro-2-methylphenylboronic acid
Formula: C₇H₈BFO₂
MW: 153.95 g/mol
Melting Pt: 148…149 °C
Storage Temperature: Ambient
MDL Number: MFCD03095047
CAS Number: 163517-62-2
EINECS: 000-000-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR