(R)-(-)-2-Hexyl isothiocyanate 95%
|
Danger
|
L20066-03
L20066-03
CAAAAL20066-03EA
335.89
CAD
CAAAAL20066-03
AAAL20066-03
(R)-(-)-2-Hexyl isothiocyanate 95%
(R)-(-)-2-Hexyl isothiocyanate
Formula:
C₇H₁₃NS Density: 0.91 Storage Temperature: Ambient |
EINECS:
000-000-0 UN: 2922 ADR: "8",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...