Arsenic, plasma standard solution, Specpure®, As 10,000µg/ml, Thermo Fisher Scientific

Supplier: Thermo Fisher Scientific
Specpure®
14369-AE 14369-AD 14369-AP 14369-AP 14369-AD 14369-AE
CAAA14369-APEA 670.67 CAD
AA14369-AE AA14369-AD CAAA14369-AP AA14369-AP CAAA14369-AD CAAA14369-AE
Arsenic, plasma standard solution, Specpure®, As 10,000µg/ml, Thermo Fisher Scientific
Standards Cationic Standards Arsenic Standards
Molecular Formula: As in 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00085309
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR