Methyl green 0.1% (w/v) in aqueous solution
Supplier: Thermo Scientific Chemicals
Synonyms:
C.I. 42590
42747-AE
42747-AE
CAAA42747-AEEA
62.85
CAD
CAAA42747-AE
AA42747-AE
Methyl green 0.1% (w/v) in aqueous solution
Methyl Green
Transition interval: pH 0.2 (yellow) to pH 1.8 (blue)
Formula:
C₂₇H₃₅BrClN₃ MW: 516.95 g/mol |
MDL Number:
MFCD00036449 CAS Number: 14855-76-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...