1,6-Dibromoperfluorohexane 96%

Warning

Synonyms: Perfluoro(1,6-dibromohexane)

B22729-06 B22729-03 B22729-03 B22729-06
CAAAB22729-06EA 302.6 CAD
CAAAB22729-06 AAB22729-06 AAB22729-03 CAAAB22729-03
1,6-Dibromoperfluorohexane 96%
1,6-Dibromoperfluorohexane
Formula: C₆Br₂F₁₂
MW: 459.87
Boiling Pt: 141…142 °C
Storage Temperature: Ambient
MDL Number: MFCD00153110
CAS Number: 918-22-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR