4-Chloroanthranilic acid

Supplier: TCI America

Synonyms: 2-Amino-4-chlorobenzoic acid

A0661-25G
TCA0661-025GEA 64.78 CAD
TCA0661-025G
4-Chloroanthranilic acid
4-Chloroanthranilic acid

CAS Number: 89-77-0
MDL Number: MFCD00007778
Molecular Formula: C7H6ClNO2
Molecular Weight: 171.58
Form: Crystal
Color: White


Other Product Names / Synonyms: 2-Amino-4-chlorobenzoic Acid

Formula: C₇H₆ClNO₂
MW: 171.58 g/mol
Boiling Pt: 330 °C (1013 hPa)
Melting Pt: 240 °C
MDL Number: MFCD00007778
CAS Number: 89-77-0
https://ca.vwr.com/stibo/web/std.lang.all/81/32/28928132.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR