2-Acetyl-5-methylfuran ≥98.0%

Supplier: TCI America

Synonyms: 1-(5-Methylfuran-2-yl)ethanone

A0983-25G
TCA0983-025GEA 79 CAD
TCA0983-025G
2-Acetyl-5-methylfuran ≥98.0%
2-Acetyl-5-methylfuran

CAS Number: 1193-79-9
MDL Number: MFCD00003243
Molecular Formula: C7H8O2
Molecular Weight: 124.14
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 69
Flash Point (°C): 79
Specific Gravity (20/20): 1.07

Formula: C₇H₈O₂
MW: 124.14 g/mol
Boiling Pt: 69 °C
Density: 1.07 (20 °C)
Flash Pt: 79 °C
MDL Number: MFCD00003243
CAS Number: 1193-79-9
https://ca.vwr.com/stibo/web/std.lang.all/87/33/28928733.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR