2-Amino-4-methylthiazole ≥99.0%

Supplier: TCI America

Synonyms: 2-Amino-4-methylthiazol4-Methylthiazol-2-ylamine

A1303-25G A1303-250G
TCA1303-250GEA 216.46 CAD
TCA1303-250G TCA1303-025G
2-Amino-4-methylthiazole ≥99.0%
2-Amino-4-methylthiazole

CAS Number: 1603-91-4
MDL Number: MFCD00005329
Molecular Formula: C4H6N2S
Molecular Weight: 114.17
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 232
Melting point (°C): 45
Flash Point (°C): 110

Formula: C₄H₆N₂S
MW: 114.17 g/mol
Boiling Pt: 232 °C
Melting Pt: 45 °C
Density: 1.202 g/cm³ (50 °C)
Flash Pt: 110 °C
MDL Number: MFCD00005329
CAS Number: 1603-91-4
Merck Index: 13,00449
https://ca.vwr.com/stibo/web/std.lang.all/92/01/28929201.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR