Cyclohexyl butyrate ≥98.0%

Supplier: TCI America

B0758-25ML
TCB0758-025MLEA 25.28 CAD
TCB0758-025ML
Cyclohexyl butyrate ≥98.0%
Cyclohexyl butyrate

CAS Number: 1551-44-6
MDL Number: MFCD00046354
Molecular Formula: C10H18O2
Molecular Weight: 170.25
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 208
Flash Point (°C): 78
Specific Gravity (20/20): 0.94


Other Product Names / Synonyms: Butyric Acid Cyclohexyl Ester

Formula: C₁₀H₁₈O₂
Boiling Pt: 208 °C
Density: 0.94 (20 °C)
Flash Pt: 78 °C
MDL Number: MFCD00046354
CAS Number: 1551-44-6

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR