Cyclooctanone ≥97.0%

Supplier: TCI America

C0504-10G C0504-25G
TCC0504-010GEA 29.55 CAD
TCC0504-010G TCC0504-025G
Cyclooctanone ≥97.0%
Cyclooctanone

CAS Number: 502-49-8
MDL Number: MFCD00001754
Molecular Formula: C8H14O
Molecular Weight: 126.20
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 45
Flash Point (°C): 74

Formula: C₈H₁₄O
MW: 126.2 g/mol
Boiling Pt: 196 °C (1013 hPa)
Melting Pt: 45 °C
Density: 0.9581 g/cm³ (20 °C)
Flash Pt: 74 °C
MDL Number: MFCD00001754
CAS Number: 502-49-8
UN: 1759
ADR: 8,III
https://ca.vwr.com/stibo/web/std.lang.all/75/33/28937533.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR