2-Chloro-1-methylpyridinium iodide ≥98.0% (by titrimetric analysis)

Supplier: TCI America

C0903-25G
TCC0903-025GEA 146.94 CAD
TCC0903-025G
2-Chloro-1-methylpyridinium iodide ≥98.0% (by titrimetric analysis)
2-Chloro-1-methylpyridinium iodide

CAS Number: 14338-32-0
MDL Number: MFCD00011984
Molecular Formula: C6H7ClIN
Molecular Weight: 255.48
Purity/Analysis Method: >98.0% (T)
Form: Crystal

Formula: C₆H₇ClIN
Melting Pt: 207 °C
MDL Number: MFCD00011984
CAS Number: 14338-32-0
https://ca.vwr.com/stibo/web/std.lang.all/81/16/28938116.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR