Ethyl-3-chloropropionate ≥97.0%

Supplier: TCI America

C1274-25G C1274-100G
TCC1274-100GEA 565.64 CAD
TCC1274-100G TCC1274-025G
Ethyl-3-chloropropionate ≥97.0%
Ethyl 3-chloropropionate

CAS Number: 623-71-2
MDL Number: MFCD00000989
Molecular Formula: C5H9ClO2
Molecular Weight: 136.58
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 162
Flash Point (°C): 54
Specific Gravity (20/20): 1.10


Other Product Names / Synonyms: 3-Chloropropionic Acid Ethyl Ester

Formula: C₅H₉ClO₂
Boiling Pt: 162 °C
Density: 1.10 (20 °C)
Flash Pt: 54 °C
MDL Number: MFCD00000989
CAS Number: 623-71-2
https://ca.vwr.com/stibo/web/std.lang.all/85/17/28938517.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR