1,2-Dibromobutane ≥98.0%

Supplier: TCI America

D0174-25G
TCD0174-025GEA 28.44 CAD
TCD0174-025G
1,2-Dibromobutane ≥98.0%
1,2-Dibromobutane

CAS Number: 533-98-2
MDL Number: MFCD00000157
Molecular Formula: C4H8Br2
Molecular Weight: 215.92
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 166
Specific Gravity (20/20): 1.80


Other Product Names / Synonyms: 1,2-Butylene Bromide

Formula: BrH₂CBrCHCH₂CH₃
MW: 215.92 g/mol
Boiling Pt: 166 °C
Melting Pt: –64 °C
Density: 1.80 (20 °C)
MDL Number: MFCD00000157
CAS Number: 533-98-2
Merck Index: 13,01565
https://ca.vwr.com/stibo/web/std.lang.all/45/72/28974572.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR