2,5-Diethoxyaniline ≥98.0% (by GC, titration analysis)

Supplier: TCI America

D0450-25G
TCD0450-025GEA 140.62 CAD
TCD0450-025G
2,5-Diethoxyaniline ≥98.0% (by GC, titration analysis)
2,5-Diethoxyaniline

CAS Number: 94-85-9
MDL Number: MFCD00015144
Molecular Formula: C10H15NO2
Molecular Weight: 181.24
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 85


Other Product Names / Synonyms: Aminohydroquinone Diethyl Ether

Formula: C₁₀H₁₅NO₂
MW: 181.23 g/mol
Boiling Pt: 190…192 °C (30 mmHg)
Melting Pt: 85 °C
MDL Number: MFCD00015144
CAS Number: 94-85-9
UN: 2811
ADR: 6.1,III
https://ca.vwr.com/stibo/web/std.lang.all/50/03/28975003.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR