1,3-Diiodopropane ≥97.0% stabilized

Supplier: TCI America

Synonyms: Trimethylene diiodide

Stabilization: Stabilised with Stabilised

D0611-25G
TCD0611-025GEA 169.06 CAD
TCD0611-025G
1,3-Diiodopropane ≥97.0% stabilized
1,3-Diiodopropane

(stabilized with Copper chip)
CAS Number: 627-31-6
MDL Number: MFCD00001097
Molecular Formula: C3H6I2
Molecular Weight: 295.89
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 111
Flash Point (°C): 110
Specific Gravity (20/20): 2.58


Other Product Names / Synonyms: Trimethylene Diiodide, Trimethylene Iodide

Formula: C₃H₆I₂
MW: 295.89 g/mol
Boiling Pt: 111 °C
Melting Pt: –20 °C
Density: 2.58 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00001097
CAS Number: 627-31-6
https://ca.vwr.com/stibo/web/std.lang.all/53/96/28975396.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR