2,2-Dimethylglutaric acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 2,2-Dimethyl-1,5-pentanedioic acid

D1786-25G
TCD1786-025GEA 715.74 CAD
TCD1786-025G
2,2-Dimethylglutaric acid ≥98.0% (by GC, titration analysis)
2,2-Dimethylglutaric acid

CAS Number: 681-57-2
MDL Number: MFCD00004197
Molecular Formula: C7H12O4
Molecular Weight: 160.17
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 200
Melting point (°C): 86


Other Product Names / Synonyms: 2,2-Dimethylpentanedioic Acid

Formula: C₇H₁₂O₄
MW: 160.17 g/mol
Boiling Pt: 200 °C
Melting Pt: 86 °C
MDL Number: MFCD00004197
CAS Number: 681-57-2
https://ca.vwr.com/stibo/web/std.lang.all/85/60/28978560.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR