4-Ethoxybenzyl alcohol ≥97.0%

Supplier: TCI America

Synonyms: (4-Ethoxyphenyl)methanol

E0492-25G
TCE0492-025GEA 557.74 CAD
TCE0492-025G
4-Ethoxybenzyl alcohol ≥97.0%
4-Ethoxybenzyl alcohol

CAS Number: 6214-44-4
MDL Number: MFCD00004655
Molecular Formula: C9H12O2
Molecular Weight: 152.19
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Flash Point (°C): 113
Freezing point (°C): 29

Formula: C₉H₁₂O₂
MW: 152.19 g/mol
Boiling Pt: 273 °C (1013 hPa)
Melting Pt: 27 °C
Flash Pt: 113 °C
MDL Number: MFCD00004655
CAS Number: 6214-44-4
https://ca.vwr.com/stibo/web/std.lang.all/80/06/28968006.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR