2-Fluorobenzonitrile ≥98.0%

Supplier: TCI America

Synonyms: o-Fluorobenzonitrile

F0220-25G
TCF0220-025GEA 85.32 CAD
TCF0220-025G
2-Fluorobenzonitrile ≥98.0%
2-Fluorobenzonitrile

CAS Number: 394-47-8
MDL Number: MFCD00001773
Molecular Formula: C7H4FN
Molecular Weight: 121.11
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 103
Flash Point (°C): 74
Specific Gravity (20/20): 1.14

Formula: C₇H₄FN
MW: 121.11 g/mol
Boiling Pt: 103 °C
Melting Pt: 35…37 °C
Density: 1.14 (20 °C)
Flash Pt: 74 °C
MDL Number: MFCD00001773
CAS Number: 394-47-8
UN: 3276
ADR: 6.1,III
https://ca.vwr.com/stibo/web/std.lang.all/88/26/28968826.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR