3-Fluoro-p-toluonitrile ≥98.0%

Supplier: TCI America

Synonyms: 3-Fluoro-4-methylbenzonitrile

F0424-5G
TCF0424-005GEA 85.32 CAD
TCF0424-005G
3-Fluoro-p-toluonitrile ≥98.0%
3-Fluoro-p-toluonitrile

CAS Number: 170572-49-3
MDL Number: MFCD00153174
Molecular Formula: C8H6FN
Molecular Weight: 135.14
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 48
Flash Point (°C): 83


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 3-Fluoro-4-methylbenzonitrile

Formula: C₈H₆FN
MW: 135.14 g/mol
Melting Pt: 48 °C
Flash Pt: 83 °C
MDL Number: MFCD00153174
CAS Number: 170572-49-3
UN: 3439
ADR: 6.1,III
https://ca.vwr.com/stibo/web/std.lang.all/90/18/28969018.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR