2-Hydroxy-5-nitropyridine ≥97.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 5-Nitro-2-pyridinol

H0673-25G H0673-250G
TCH0673-025GEA 216.46 CAD
TCH0673-025G TCH0673-250G
2-Hydroxy-5-nitropyridine ≥97.0% (by GC, titration analysis)
2-Hydroxy-5-nitropyridine

CAS Number: 5418-51-9
MDL Number: MFCD00006276
Molecular Formula: C5H4N2O3
Molecular Weight: 140.10
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Melting point (°C): 186

Formula: C₅H₄N₂O₃
MW: 140.1 g/mol
Boiling Pt: 180…200 °C (0.2 mmHg)
Melting Pt: 186 °C
MDL Number: MFCD00006276
CAS Number: 5418-51-9
https://ca.vwr.com/stibo/web/std.lang.all/06/91/28970691.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR