4-Hydroxy-3,5-dimethylbenzoic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

H0938-5G
TCH0938-005GEA 178.54 CAD
TCH0938-005G
4-Hydroxy-3,5-dimethylbenzoic acid ≥98.0% (by titrimetric analysis)
4-Hydroxy-3,5-dimethylbenzoic acid

CAS Number: 4919-37-3
MDL Number: MFCD00016536
Molecular Formula: C9H10O3
Molecular Weight: 166.18
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 225

Formula: C₉H₁₀O₃
MW: 166.18 g/mol
Melting Pt: 225 °C
MDL Number: MFCD00016536
CAS Number: 4919-37-3
https://ca.vwr.com/stibo/web/std.lang.all/09/58/28970958.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR