Isophthalaldehyde ≥98.0%

Supplier: TCI America

Synonyms: IsophthaldialdehydeIsophthaldicarboxaldehydem-Phthalaldehyde

I0153-250G
TCI0153-250GEA 388.68 CAD
TCI0153-250G
Isophthalaldehyde ≥98.0%
Isophthalaldehyde

CAS Number: 626-19-7
MDL Number: MFCD00003372
Molecular Formula: C8H6O2
Molecular Weight: 134.13
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 136
Melting point (°C): 89


Product Categories: Synthetic Organic Chemistry, Aldehydes (Multi-component Reaction)
Other Product Names / Synonyms: Isophthaldialdehyde, Benzene-1,3-dicarboxaldehyde

Formula: C₈H₆O₂
MW: 134.13 g/mol
Boiling Pt: 136 °C
Melting Pt: 89 °C
MDL Number: MFCD00003372
CAS Number: 626-19-7
https://ca.vwr.com/stibo/web/std.lang.all/19/34/28971934.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR