1-Nitroso-2-naphthol ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 2-Hydroxy-1-nitrosonaphthalene1-Nitrosonaphthalen-2-ol

N0266-25G
TCN0266-025GEA 88.48 CAD
TCN0266-025G
1-Nitroso-2-naphthol ≥98.0% (by GC, titration analysis)
1-Nitroso-2-naphthol

CAS Number: 131-91-9
MDL Number: MFCD00003884
Molecular Formula: C10H7NO2
Molecular Weight: 173.17
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 107

Formula: C₁₀H₇NO₂
MW: 173.17 g/mol
Boiling Pt: 304 °C (1013 hPa)
Melting Pt: 107 °C
MDL Number: MFCD00003884
CAS Number: 131-91-9
Merck Index: 12,06739
https://ca.vwr.com/stibo/web/std.lang.all/93/48/28989348.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR