trans-2-Nonenal ≥95.0%
Supplier: TCI America
Synonyms:
(E)-2-Nonenal
N0483-25ML
N0483-5ML
TCN0483-005MLEA
76.47
CAD
TCN0483-005ML
TCN0483-025ML
trans-2-Nonenal ≥95.0%
trans-2-Nonenal
CAS Number: 18829-56-6
MDL Number: MFCD00007012
Molecular Formula: C9H16O
Molecular Weight: 140.23
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 108
Flash Point (°C): 84
Specific Gravity (20/20): 0.85
Formula:
C₉H₁₆O MW: 140.23 g/mol Boiling Pt: 108 °C Density: 0.85 (20 °C) Flash Pt: 84 °C |
MDL Number:
MFCD00007012 CAS Number: 18829-56-6 Merck Index: 12,06771 |
![]() |
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...