Phenethylamine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: β-Phenylethylamine2-PhenylethylaminePEA

P0085-500ML P0085-25ML P0085-100ML
TCP0085-100MLEA 69.52 CAD
TCP0085-500ML TCP0085-25ML TCP0085-100ML
Phenethylamine ≥98.0% (by GC, titration analysis)
Phenethylamine

CAS Number: 64-04-0
MDL Number: MFCD00008184
Molecular Formula: C8H11N
Molecular Weight: 121.18
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Flash Point (°C): 90
Specific Gravity (20/20): 0.96


Product Categories: Synthetic Organic Chemistry, Amines (Multi-component Reaction)
Other Product Names / Synonyms: 2-Aminoethylbenzene, 1-Amino-2-phenylethane, Phenethylamine

Formula: C₆H₅CH₂CH₂NH₂
MW: 121.18 g/mol
Boiling Pt: 200 °C (1013 hPa)
Melting Pt: –60 °C
Density: 0.96 (20 °C)
Flash Pt: 90 °C
MDL Number: MFCD00008184
CAS Number: 64-04-0
UN: 2735
ADR: 8,I
Merck Index: 12,07371
https://ca.vwr.com/stibo/web/std.lang.all/07/83/28990783.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR