(±)-4-Phenyl-2-butanol ≥99.0%

Supplier: TCI America

P0876-25ML
TCP0876-025MLEA 246.48 CAD
TCP0876-025ML
(±)-4-Phenyl-2-butanol ≥99.0%
(±)-4-Phenyl-2-butanol

CAS Number: 2344-70-9
MDL Number: MFCD00044349
Molecular Formula: C10H14O
Molecular Weight: 150.22
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 239
Flash Point (°C): 110
Specific Gravity (20/20): 0.98

Formula: (C₆H₅)CH₂CH₂CH(OH)CH₃
MW: 150.22 g/mol
Boiling Pt: 239 °C
Density: 0.98 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00044349
CAS Number: 2344-70-9
https://ca.vwr.com/stibo/web/std.lang.all/15/44/28991544.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR