1,2-Phenylenediacetic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 2,2'-(1,2-Phenylene)diacetic acid

P1052-5G
TCP1052-005GEA 48.98 CAD
TCP1052-005G
1,2-Phenylenediacetic acid ≥98.0% (by HPLC, titration analysis)
1,2-Phenylenediacetic acid

CAS Number: 7500-53-0
MDL Number: MFCD00004329
Molecular Formula: C10H10O4
Molecular Weight: 194.19
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: Very Pale Yellow
Melting point (°C): 150

Formula: C₁₀H₁₀O₄
MW: 194.19 g/mol
Melting Pt: 150 °C
MDL Number: MFCD00004329
CAS Number: 7500-53-0
https://ca.vwr.com/stibo/web/std.lang.all/16/87/28991687.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR