Quinazoline ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Q0055-1G
TCQ0055-001GEA 186.44 CAD
TCQ0055-001G
Quinazoline ≥98.0% (by GC, titration analysis)
Quinazoline

CAS Number: 253-82-7
MDL Number: MFCD00006712
Molecular Formula: C8H6N2
Molecular Weight: 130.15
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Boiling point (°C): 243
Melting point (°C): 48
Flash Point (°C): 106

Formula: C₈H₆N₂
Boiling Pt: 243 °C
Melting Pt: 48 °C
Flash Pt: 106 °C
MDL Number: MFCD00006712
CAS Number: 253-82-7
https://ca.vwr.com/stibo/web/std.lang.all/37/01/28993701.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR