Succinimide ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Pyrrolidine-2,5-dione

S0108-500G S0108-25G
TCS0108-500GEA 102.7 CAD
TCS0108-500G TCS0108-025G
Succinimide ≥98.0% (by titrimetric analysis)
Succinimide

CAS Number: 123-56-8
MDL Number: MFCD00005495
Molecular Formula: C4H5NO2
Molecular Weight: 99.09
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Boiling point (°C): 130
Melting point (°C): 125
Flash Point (°C): 201


Other Product Names / Synonyms: 2,5-Pyrrolidinedione

Formula: C₄H₅NO₂
MW: 99.09 g/mol
Boiling Pt: 130 °C
Melting Pt: 125 °C
Density: 1.418 g/cm³ (20 °C)
Flash Pt: 201 °C
MDL Number: MFCD00005495
CAS Number: 123-56-8
Merck Index: 12,09040
https://ca.vwr.com/stibo/web/std.lang.all/39/54/28993954.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR