n-Pentadecane ≥99.5%

Supplier: TCI America

Synonyms: N-PentadecanePentadecane

S0287-5ML
TCS0287-005MLEA 116.92 CAD
TCS0287-005ML
n-Pentadecane ≥99.5%
n-Pentadecane

[Standard Material for GC]
CAS Number: 629-62-9
MDL Number: MFCD00008990
Molecular Formula: C15H32
Molecular Weight: 212.42
Purity/Analysis Method: >99.5% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 271
Flash Point (°C): 120
Freezing point (°C): 10
Specific Gravity (20/20): 0.77

Formula: H₃C(CH₂)₁₃CH₃
MW: 212.42 g/mol
Boiling Pt: 271 °C
Melting Pt: 13…15 °C
Density: 0.77 (20 °C)
Flash Pt: 120 °C
MDL Number: MFCD00008990
CAS Number: 629-62-9
UN: 3295
ADR: 3,III

Order Now

Specification Test Results

Purity/Analysis Method 99.5% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR