Methyl myristate ≥99.5%

Supplier: TCI America

Synonyms: Methyl tetradecanoateTetradecylic acid methyl esterMyristic acid methyl esterTetradecanoic acid methyl ester

S0310-5ML
TCS0310-005MLEA 206.82 CAD
TCS0310-005ML
Methyl myristate ≥99.5%
Methyl myristate

[Standard Material for GC]
CAS Number: 124-10-7
MDL Number: MFCD00008983
Molecular Formula: C15H30O2
Molecular Weight: 242.40
Purity/Analysis Method: >99.5% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 173
Flash Point (°C): 157
Freezing point (°C): 18
Specific Gravity (20/20): 0.87


Product Categories: Analytical Science, Fatty Acid Methyl Esters (GC Standard)
Other Product Names / Synonyms: Methyl Tetradecanoate, Myristic Acid Methyl Ester, Tetradecanoic Acid Methyl Ester

Formula: C₁₅H₃₀O₂
MW: 242.4 g/mol
Boiling Pt: 173 °C
Melting Pt: 18 °C
Density: 0.87 (20 °C)
Flash Pt: 157 °C
MDL Number: MFCD00008983
CAS Number: 124-10-7

Order Now

Specification Test Results

Purity/Analysis Method 99.5% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR