1-Tetradecene ≥99.5%

Supplier: TCI America

S0344-5ML
TCS0344-005MLEA 150.1 CAD
TCS0344-005ML
1-Tetradecene ≥99.5%
1-Tetradecene

[Standard Material for GC]
CAS Number: 1120-36-1
MDL Number: MFCD00008981
Molecular Formula: C14H28
Molecular Weight: 196.38
Purity/Analysis Method: >99.5% (GC)
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 112
Specific Gravity (20/20): 0.77

Formula: C₁₄H₂₈
Boiling Pt: 251 °C (1013 hPa)
Melting Pt: –13 °C
Density: 0.77 (20 °C)
Flash Pt: 112 °C
MDL Number: MFCD00008981
CAS Number: 1120-36-1

Order Now

Specification Test Results

Purity/Analysis Method 99.5% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR