Tetraallyloxyethane ≥90.0%

Synonyms: 1,1,2,2-Tetrakis(allyloxy)ethane

T0030-25ML
TCT0030-025MLEA 30.13 CAD
TCT0030-025ML
Tetraallyloxyethane ≥90.0%
Tetraallyloxyethane

CAS Number: 16646-44-9
MDL Number: MFCD00043927
Molecular Formula: C14H22O4
Molecular Weight: 254.33
Purity/Analysis Method: >90.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 160
Flash Point (°C): 113
Specific Gravity (20/20): 0.97


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 1,1,2,2-Tetrakis(allyloxy)ethane

Formula: C₁₄H₂₂O₄
MW: 254.33 g/mol
Boiling Pt: 160 °C
Density: 0.97 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00043927
CAS Number: 16646-44-9
https://ca.vwr.com/stibo/web/std.lang.all/43/74/28994374.jpg

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR