1,2,4,5-Tetramethylbenzene ≥99.8%, zone refined: number of passes 23

Supplier: TCI America

T0140-ZONE
TCT0140-ZONEEA 263.86 CAD
TCT0140-ZONE
1,2,4,5-Tetramethylbenzene ≥99.8%, zone refined: number of passes 23
1,2,4,5-Tetramethylbenzene

CAS Number: 95-93-2
MDL Number: MFCD00008528
Molecular Formula: C10H14
Molecular Weight: 134.22
Purity/Analysis Method: >99.8% (GC)
Form: Crystal
Color: White
Boiling point (°C): 197
Melting point (°C): 80
Flash Point (°C): 73


Product Categories: Materials Chemistry, Highly Purified Reagents
Other Product Names / Synonyms: Durene

Formula: C₁₀H₁₄
MW: 134.22 g/mol
Boiling Pt: 197 °C
Melting Pt: 80 °C
Density: 1.036 g/cm³ (20 °C)
Flash Pt: 73 °C
MDL Number: MFCD00008528
CAS Number: 95-93-2
UN: 1325
ADR: 4.1,II
Merck Index: 13,03501
https://ca.vwr.com/stibo/web/std.lang.all/44/64/28994464.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.8% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR