Tetrapropylammonium iodide ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Tetra-n-propylammonium iodide

T0172-25G
TCT0172-025GEA 133.83 CAD
TCT0172-025G
Tetrapropylammonium iodide ≥98.0% (by titrimetric analysis)
Tetrapropylammonium iodide

CAS Number: 631-40-3
MDL Number: MFCD00011842
Molecular Formula: C12H28IN
Molecular Weight: 313.27
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 285

Formula: C₁₂H₂₈IN
Melting Pt: 285 °C
Density: 1.317 g/cm³ (20 °C)
MDL Number: MFCD00011842
CAS Number: 631-40-3
https://ca.vwr.com/stibo/web/std.lang.all/44/92/28994492.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR