Chlorotriethylgermane ≥97.0%

Supplier: TCI America

Synonyms: TriethylchlorogermaneTriethylgermanium chlorideChlorotriethylgermane

T1160-1G
TCT1160-001GEA 285.98 CAD
TCT1160-001G
Chlorotriethylgermane ≥97.0%
Chlorotriethylgermane

CAS Number: 994-28-5
MDL Number: MFCD00013591
Molecular Formula: C6H15ClGe
Molecular Weight: 195.27
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 176
Flash Point (°C): 110
Specific Gravity (20/20): 1.16


Other Product Names / Synonyms: Triethylgermanium Chloride

Formula: C₆H₁₅ClGe
Boiling Pt: 176 °C
Density: 1.16 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00013591
CAS Number: 994-28-5
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR