Tetraheptylammonium iodide ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: TetraheptylammoniumiodideTetra-n-heptylammonium iodide

T1396-25G
TCT1396-025GEA 146.94 CAD
TCT1396-025G
Tetraheptylammonium iodide ≥98.0% (by titrimetric analysis)
Tetraheptylammonium iodide

CAS Number: 3535-83-9
MDL Number: MFCD00041984
Molecular Formula: C28H60IN
Molecular Weight: 537.70
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 123

Formula: C₂₈H₆₀IN
MW: 537.7 g/mol
Melting Pt: 123 °C
Density: 1.066 g/cm³ (25 °C)
MDL Number: MFCD00041984
CAS Number: 3535-83-9
https://ca.vwr.com/stibo/web/std.lang.all/54/18/28995418.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR