3,3,3-Triphenylpropionic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 3,3,3-Triphenylpropanoic acid

T1558-25G T1558-5G
TCT1558-25GEA 565.64 CAD
TCT1558-005G TCT1558-25G
3,3,3-Triphenylpropionic acid ≥98.0% (by GC, titration analysis)
3,3,3-Triphenylpropionic acid

CAS Number: 900-91-4
MDL Number: MFCD00002713
Molecular Formula: C21H18O2
Molecular Weight: 302.37
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 181

Formula: C₂₁H₁₈O₂
MW: 302.37 g/mol
Melting Pt: 181 °C
Density: 1.264 g/cm³ (20 °C)
MDL Number: MFCD00002713
CAS Number: 900-91-4
https://ca.vwr.com/stibo/web/std.lang.all/55/69/28995569.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR