Methyl undecanoate ≥98.0%

Supplier: TCI America

Synonyms: Undecanoic acid methyl esterUndecylic acid methyl ester

U0050-25ML
TCU0050-025MLEA 142.2 CAD
TCU0050-025ML
Methyl undecanoate ≥98.0%
Methyl undecanoate

CAS Number: 1731-86-8
MDL Number: MFCD00008957
Molecular Formula: C12H24O2
Molecular Weight: 200.32
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 123
Melting point (°C): -10
Flash Point (°C): 109
Specific Gravity (20/20): 0.87


Product Categories: Bioscience, Lipids
Other Product Names / Synonyms: Undecanoic Acid Methyl Ester

Formula: C₁₂H₂₄O₂
Boiling Pt: 123 °C
Melting Pt: –10 °C
Density: 0.87 (20 °C)
Flash Pt: 109 °C
MDL Number: MFCD00008957
CAS Number: 1731-86-8
https://ca.vwr.com/stibo/web/std.lang.all/72/11/28997211.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR