- Home
- Pumps
- Vacuum Aspiration Systems
- VACUUBRAND® BVC Biofluid Aspiration Systems, BrandTech
VACUUBRAND® BVC Biofluid Aspiration Systems, BrandTech
Supplier: BrandTech
20727200
20727503
20727403
20727100
20727400
20727303
20727000
20727203
20727500
20727300
89239-604EA
5733.87
CAD
89239-604
89239-618
89239-614
89239-602
89239-612
89239-610
89239-600
89239-606
89239-616
89239-608
VACUUBRAND® BVC Biofluid Aspiration Systems, BrandTech
Pumps
Vacuum Aspiration Systems
The new family of BVC fluid aspiration systems offer the perfect solution for safe, sensitive, and efficient aspiration of supernatants for a modern Biology laboratory
- Safe containment of biofluids
- Quiet, easy to use
- Powerful enough for two users
- Premium quality for long service life
VACUUBRAND® BVC systems ensure reliable aspiration capability. Control and Professional models feature powerful VACUUBRAND® chemistry-design vacuum pumps, which offer exceptionally long service life and enough power for two users. High-quality components enable years of trouble-free operation, even when working with aggressive disinfectants. This results in a low cost of operation. High-capacity 2 or 4 liter collection bottles, an autoclavable hydrophobic 0.2μm bio-filter, and an ergonomic VacuuHandControl (VHC) keep potentially biohazardous materials contained. The VHC features a functional design which keeps all fluid within the aspiration tubing. The "G" models feature a 2 liter glass collection vessel with scratch and fragment protection coating for true bleach resistance. Streamlined design incorporates bottle, controls, and pump (Control and Professional models) in a small footprint that fits easily on a lab bench or next to a Biological Safety Cabinet. Additionally, a clip is included to help secure all tubing out of the way. Smooth contours and touch panels, rather than knobs, ensure the system is easy to keep clean as well.
The BVC Basic is the unpowered version, suitable for use with a VACUULAN® vacuum network, central vacuum system, or shared pump. A mechanical regulator provides a suitable vacuum level in the bottle for aspiration, and shuts off connection to the line when appropriate vacuum level is reached. This reduces demand on the vacuum supply, increasing efficiency while reducing noise and impact on other applications. The Basic system is supplied ready for use, and includes a stable and easy-to-clean powder-coated metal stand, 0.2μm aerosol filter, and VacuuHandControl. Available in heavy duty polypropylene, or bleach resistant glass versions.
The BVC Control features a powerful whisper-quiet VACUUBRAND® chemistry-design vacuum pump, eliminating the need for a central vacuum supply. Potentially biohazardous materials can be contained in a local area. The pump is turned on and off automatically on demand when using the VacuuHandControl, which further reduces noise levels. An easy to clean touch panel adjusts and indicates the vacuum level switchpoint. This provides exactly the aspiration power needed, at all times. Available with either a 4 liter polypropylene, or 2 liter bleach-resistant glass bottle.
The top-of-the-line BVC Professional includes an external liquid level sensor, which operates without contacting or being exposed to bottle contents. This allows users to concentrate fully on work without worrying about overflow. A special disinfection program enables the aspiration of disinfectant after the system is shut down to decontaminate the VacuuHandControl. The 4L polypropylene bottle version is supplied with quick connectors as standard. The 2 liter glass bottle “bleach proof” version includes hose barb connections, although optional quick connectors are available. The contact-free design of the level sensor simplifies container design and reduces costs when using multiple collection bottles.
All models feature integrated 2 or 4 liter containment vessels for the safe collection of biological waste. An autoclavable hydrophobic 0.2µm bio-filter protects the vacuum supply and laboratory environment from biohazardous aerosols. The VacuuHandControl (VHC) provides thumb-pressure control for precise aspiration and keeps all fluid within the aspiration tubing for containment and cleaning convenience.
Certifications: The 230V models 89239-604, -608, -612, and -616 are CE marked.
Ordering information: BVC systems inlcude base, collection bottle, one VHC hand controller, operating manual, and one-year warranty.
New Product
Limited Time Sale
Select items below are currently on sale
Enter Certificate Details
Inquire for Price
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Item(s) Added to Cart